Bromkresolgrün
Lieferant: Thermo Fisher Scientific
A17090.06
A17090.14
A17090.06EA
35.6
EUR
A17090.06
A17090.14
Bromkresolgrün
Bromkresolgrün
CAS No.: 76-60-8
Molecular Formula: C21H14Br4O5S
Formula Weight: 698.02
Melting Point: ca 225° dec.
Beilstein Reference: 372527
Merck Reference: 14,1386
MDL No.: MFCD00005874
Formel:
C₂₁H₁₄Br₄O₅S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005874 CAS-Nummer: 76-60-8 EINECS: 200-972-8 Merck Index: 14,1386 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...