(±)-1,2-Dibromhexafluorpropan 95%

Lieferant: Thermo Fisher Scientific
Achtung

A17103.14
A17103.14EA 41.8 EUR
A17103.14
(±)-1,2-Dibromhexafluorpropan 95%
(±)-1,2-Dibromhexafluorpropan

Purity: 95% CAS No.: 661-95-0 Molecular Formula: BrCF2CFBrCF3 Formula Weight: 309.84 Boiling Point: 71-72° MDL No.: MFCD00013568

Formel: BrF₂CBrCFCF₃
Molecular Weight: 309.83 g/mol
Siedepunkt: 71…72 °C
Dichte: 2.169
Lagertemperatur: Raumtemperatur
MDL: MFCD00013568
CAS-Nummer: 661-95-0
EINECS: 211-550-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR