1,4-Di-tert-butylbenzol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A17146.06
A17146.14
A17146.06EA
49.1
EUR
A17146.06
A17146.14
1,4-Di-tert-butylbenzol ≥98%
1,4-Di-tert-butylbenzol
Purity: 98%
CAS No.: 1012-72-2
Molecular Formula: C14H22
Formula Weight: 190.33
Melting Point: 75-79°
Boiling Point: 236°
MDL No.: MFCD00008836
Formel:
C₁₄H₂₂ Siedepunkt: 236 °C Schmelzpunkt: 75…79 °C Dichte: 0,985 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008836 CAS-Nummer: 1012-72-2 EINECS: 213-790-9 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...