Nile blue A

Lieferant: Thermo Fisher Scientific

Synonyme: Nile blue A sulphate, Nile blue sulphate, 5-Amino-9-(diethylamino)benzo[a]phenoxazin-7-ium sulphate, Basic Blue 12

A17174.14 A17174.06
A17174.14EA 73.6 EUR
A17174.14 A17174.06
Nile blue A
Nile blue A

CAS No.: 3625-57-8 Molecular Formula: C40H40N6O6S Formula Weight: 732.84 Melting Point: 300° dec. MDL No.: MFCD00064529

Formel: C₄₀H₄₀N₆O₆S
Schmelzpunkt: >300 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064529
CAS-Nummer: 3625-57-8
EINECS: 222-832-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR