(S)-(-)-1-(1-Naphthyl)ethylamin 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99%
CAS No.: 10420-89-0
Molecular Formula: C12H13N
Formula Weight: 171.24
Storage Temperature: 2°C to 8°C
Flash Point:
Formel:
C₁₂H₁₃N Siedepunkt: 152…153 °C (11 mmHg) Dichte: 1,060 Flammpunkt: >110 °C (230 °F) |
MDL:
MFCD00064179 CAS-Nummer: 10420-89-0 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...