3,3'-Thiodipropionsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,3-Thiodipropionic acid

A17220.30 A17220.0B
A17220.30EA 29.6 EUR
A17220.30 A17220.0B
3,3'-Thiodipropionsäure ≥98%
3,3'-Thiodipropionsäure

Purity: 98% CAS No.: 111-17-1 Molecular Formula: C6H10O4S Formula Weight: 178.21 Melting Point: 127-134° Beilstein Reference: 1210299 Merck Reference: 14,9332 MDL No.: MFCD00002781

Formel: C₆H₁₀O₄S
Siedepunkt: 347 °C (1013 hPa)
Schmelzpunkt: 127…134 °C
Dichte: 1,496 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002781
CAS-Nummer: 111-17-1
EINECS: 203-841-3
Merck Index: 14,9332

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR