Kresolrot

Lieferant: Thermo Fisher Scientific
Achtung

A17243.14 A17243.22
A17243.14EA 48.2 EUR
A17243.14 A17243.22
Kresolrot
Kresolrot

CAS No.: 1733-12-6 Molecular Formula: C21H18O5S Formula Weight: 382.44 Melting Point: 300° Beilstein Reference: 343399 Merck Reference: 14,2578 MDL No.: MFCD00005878

Formel: C₂₁H₁₈O₅S
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005878
CAS-Nummer: 1733-12-6
EINECS: 217-064-2
Merck Index: 14,2578

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR