3',4'-Difluorpropiophenon ≥97%
Lieferant: Thermo Scientific
Synonyme:
1-(3,4-Difluorophenyl)propan-1-one
A17257.06
A17257.06EA
97.8
EUR
A17257.06
3',4'-Difluorpropiophenon ≥97%
3',4'-Difluorpropiophenon
Purity: 97%
CAS No.: 23384-72-7
Molecular Formula: C9H8F2O
Formula Weight: 170.16
Refractive Index: 1.4880
Boiling Point: 64-65°/1.8mm
MDL No.: MFCD00015505
Formel:
C₉H₈F₂O Molecular Weight: 170,16 g/mol Siedepunkt: 64…65 °C (1,8 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015505 CAS-Nummer: 23384-72-7 EINECS: 245-627-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...