N-Acetyl-L-tyrosin 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: N-Acetyl-L-Tyrosine, (S)-2-Acetamido-3-(4-hydroxyphenyl)propanoic acid, Ac-Tyr-OH

A17307.09 A17307.18 A17307.30
A17307.09EA 62 EUR
A17307.09 A17307.18 A17307.30
N-Acetyl-L-tyrosin 99%
N-Acetyl-L-tyrosin

Purity: 99% CAS No.: 537-55-3 Molecular Formula: C11H13NO4 Formula Weight: 223.23 Melting Point: 150-151° Beilstein Reference: 2697172 Optical Rotation: +58° (c=5 in ethanol) MDL No.: MFCD00037190

Formel: C₁₁H₁₃NO₄
Schmelzpunkt: 150…151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00037190
CAS-Nummer: 537-55-3
EINECS: 208-671-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR