Hexachlor-1,3-butadien ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
A17316.18
A17316.18EA
53.3
EUR
A17316.18
Hexachlor-1,3-butadien ≥97%
Hexachlor-1,3-butadien
Purity: 97%
CAS No.: 87-68-3
Molecular Formula: CCl2=CClCCl=CCl2
Formula Weight: 260.76
Refractive Index: 1.5550
Odour: Mild
Boiling Point: 215°
Beilstein Reference: 1766570
Merck Reference: 14,4678
UN No.: 2279
MDL No.: MFCD00000836
Formel:
C₄Cl₆ Molecular Weight: 260,76 g/mol Siedepunkt: 215 °C Schmelzpunkt: –21 °C Dichte: 1,680 Flammpunkt: 90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000836 CAS-Nummer: 87-68-3 EINECS: 201-765-5 UN: 2279 ADR: 6.1,III Merck Index: 14,4678 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...