(±)-α-Brom-γ-butyrolacton ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (±)-2-Brombutan-4-olid, (±)-2-Brom-4-butanolid, (±)-2-Brom-4-hydroxybuttersäurelacton

A17344.14 A17344.22
A17344.14EA 37.2 EUR
A17344.14 A17344.22
(±)-α-Brom-γ-butyrolacton ≥97%
(±)-α-Brom-γ-butyrolacton

Purity: 97% CAS No.: 5061-21-2 Molecular Formula: C4H5BrO2 Formula Weight: 164.99 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5080 Boiling Point: 137-139°/6mm Beilstein Reference: 107800 MDL No.: MFCD00005387

Formel: C₄H₅BrO₂
Siedepunkt: 137…139 °C (6 mmHg)
Dichte: 1,994
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00005387
CAS-Nummer: 5061-21-2
EINECS: 225-764-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR