N,N-Dimethylhexylamin 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: N,N-Dimethylhexan-1-amine

A17364.06 A17364.14 A17364.22
A17364.06EA 59.3 EUR
A17364.06 A17364.14 A17364.22
N,N-Dimethylhexylamin 99%
N,N-Dimethylhexylamin

Purity: 99% CAS No.: 4385-04-0 Molecular Formula: CH3(CH2)5N(CH3)2 Formula Weight: 129.25 Flash Point: 33°(92°F) Refractive Index: 1.4136 Boiling Point: 146-150° UN No.: 2733 MDL No.: MFCD00009522

Formel: C₈H₁₉N
Molecular Weight: 129.25 g/mol
Siedepunkt: 146…150 °C
Dichte: 0.744
Flammpunkt: 33 °C (92 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009522
CAS-Nummer: 4385-04-0
EINECS: 224-491-8
UN: 2733
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR