EDTA Tetranatriumsalz Hydrat 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A17385.0B
A17385.0I
A17385.30
A17385.0BEA
59.5
EUR
A17385.0B
A17385.0I
A17385.30
EDTA Tetranatriumsalz Hydrat 98%
EDTA Tetranatriumsalz Hydrat
Purity: 98%
CAS No.: 194491-31-1
Molecular Formula: C10H12N2Na4O8.xH2O
Formula Weight: 380.17(anhy)
Melting Point:
Formel:
C₁₀H₁₄N₂Na₄O₉ Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150025 CAS-Nummer: 194491-31-1 EINECS: 200-573-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...