Chlorphenesin ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A17386.14
A17386.22
A17386.36
A17386.14EA
45
EUR
A17386.14
A17386.22
A17386.36
Chlorphenesin ≥99%
Chlorphenesin
Purity: 99%
CAS No.: 104-29-0
Molecular Formula: C9H11ClO3
Formula Weight: 202.64
Melting Point: 78-82°
MDL No.: MFCD00021990
Formel:
C₉H₁₁ClO₃ Molecular Weight: 202.64 g/mol Schmelzpunkt: 78…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00021990 CAS-Nummer: 104-29-0 EINECS: 203-192-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...