Methylrot Natriumsalz

Lieferant: Thermo Fisher Scientific

A17455.14 A17455.22 A17455.36
A17455.14EA 25.6 EUR
A17455.14 A17455.22 A17455.36
Methylrot Natriumsalz
Methylrot Natriumsalz

CAS No.: 845-10-3 Molecular Formula: C15H14N3NaO2 Formula Weight: 291.29 Beilstein Reference: 4065488 MDL No.: MFCD00002426

Formel: C₁₅H₁₄N₃NaO₂
Schmelzpunkt: > 180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002426
CAS-Nummer: 845-10-3
EINECS: 212-682-9
Merck Index: 13,06142

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR