Ethylenglycolmonomethylether (2-Methoxyethanol) ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Methylglycol, Ethylenglycolmonomethylether

A17459.0D A17459.0F A17459.AP
A17459.0DEA 296 EUR
A17459.0D A17459.0F A17459.AP
Ethylenglycolmonomethylether (2-Methoxyethanol) ≥99%
Ethylenglycolmonomethylether (2-Methoxyethanol)

Purity: 99% CAS No.: 109-86-4 Molecular Formula: C3H8O2 Formula Weight: 76.10 Flash Point: 46°(115°F) Refractive Index: 1.4025 Odour: Characteristic pH: 4-7 Melting Point: -85° Boiling Point: 124-125° Beilstein Reference: 1731074 Merck Reference: 14,6038 UN No.: 1188 MDL No.: MFCD00002867

Formel: C₃H₈O₂
Siedepunkt: 124…125 °C
Schmelzpunkt: –85 °C
Dichte: 0,965
Flammpunkt: 46 °C (115 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002867
CAS-Nummer: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III
Merck Index: 14,6038

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR