Natriumcarbonat-Decahydrat ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Soda
A17484.36
A17484.0E
A17484.0C
A17484.36EA
24.6
EUR
A17484.36
A17484.0E
A17484.0C
Natriumcarbonat-Decahydrat ≥99%
Natriumcarbonat-Decahydrat
Purity: 99+%
CAS No.: 6132-02-1
Molecular Formula: CNa2O3.10H2O
Formula Weight: 286.14 (105.99anhy)
Melting Point: 32-34°
Merck Reference: 14,8596
MDL No.: MFCD00149178
Formel:
Na₂CO₃·10H₂O Molecular Weight: 286.14 g/mol Schmelzpunkt: 32…34 °C Dichte: 1.46 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149178 CAS-Nummer: 6132-02-1 EINECS: 207-838-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...