Procain Hydrochlorid 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Diethylaminoethyl 4-aminobenzoate hydrochloride
A17485.18
A17485.30
A17485.18EA
31.8
EUR
A17485.18
A17485.30
Procain Hydrochlorid 99%
Procain Hydrochlorid
Purity: 99%
CAS No.: 51-05-8
Molecular Formula: C13H20N2O2.HCl
Formula Weight: 272.78
Odour: Odourless
Melting Point: 155-157°
Boiling Point: 195-196°/17mm
Merck Reference: 14,7757
UN No.: 2811
MDL No.: MFCD00013000
Formel:
C₁₃H₂₁ClN₂O₂ Molecular Weight: 272.77 g/mol Siedepunkt: 195…196 °C (17 mmHg) Schmelzpunkt: 155…157 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013000 CAS-Nummer: 51-05-8 EINECS: 200-077-2 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...