6-Brompiperonal ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
6-Brom-1,3-benzodioxol-5-carbaldehyd
A17510.06
A17510.14
A17510.22
A17510.06EA
59.9
EUR
A17510.06
A17510.14
A17510.22
6-Brompiperonal ≥98%
6-Brompiperonal
Purity: 98%
CAS No.: 15930-53-7
Molecular Formula: C8H5BrO3
Formula Weight: 229.03
Melting Point: 127-130°
MDL No.: MFCD00022952
Formel:
C₈H₅BrO₃ Schmelzpunkt: 127…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022952 CAS-Nummer: 15930-53-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...