3-Chlor-4-hydroxybenzaldehyd 98

Lieferant: Thermo Fisher Scientific
Achtung

A17515.14 A17515.06
A17515.14EA 322 EUR
A17515.14 A17515.06
3-Chlor-4-hydroxybenzaldehyd 98
3-Chloro-4-hydroxybenzaldehyde

Purity: 98% CAS No.: 2420-16-8 Molecular Formula: C7H5ClO2 Formula Weight: 156.57 Melting Point: 128-132° MDL No.: MFCD00016981

Formel: C₇H₅ClO₂
Molecular Weight: 156.57 g/mol
Siedepunkt: 251
Schmelzpunkt: 128…132 °C
Dichte: 1.0404 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00016981
CAS-Nummer: 2420-16-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR