Dicinnamylidenaceton ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,9-Diphenylnona-1,3,6,8-tetraen-5-one, 1,9-Diphenyl-1,3,6,8-nonatetraen-5-one
A17568.06
A17568.06EA
31.4
EUR
A17568.06
Dicinnamylidenaceton ≥97%
Dicinnamalacetone
Purity: 98%
CAS No.: 622-21-9
Molecular Formula: C21H18O
Formula Weight: 286.37
Melting Point: 141-143°
MDL No.: MFCD00004792
Formel:
C₂₁H₁₈O Schmelzpunkt: 141…143 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004792 CAS-Nummer: 622-21-9 EINECS: 210-724-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...