Dexamethason ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
9-Fluor-11β,17α,21-trihydroxy-16α-methyl-pregna-1,4-dien-3,20-dion, Dexamethasonum
A17590.06
A17590.03
A17590.14
A17590.06EA
418
EUR
A17590.06
A17590.03
A17590.14
Dexamethason ≥98%
Dexamethason
Purity: 98%
CAS No.: 50-02-2
Molecular Formula: C22H29FO5
Formula Weight: 392.47
Melting Point: ca 260° dec.
Beilstein Reference: 2066651
Merck Reference: 14,2943
Optical Rotation: +75° (c=1 in doxane)
MDL No.: MFCD00064136
Anti-inflammatory glucocorticoid
Formel:
C₂₂H₂₉FO₅ Schmelzpunkt: ∼ 260 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064136 CAS-Nummer: 50-02-2 EINECS: 200-003-9 Merck Index: 14,2943 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...