Adenin Hydrochlorid, (max. 5% H₂O) ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Purin-6-amin Hydrochlorid, 1H-Adenin Hydrochlorid, 9H-Purin-6-amin Hydrochlorid, 6-Aminopurin Hydrochlorid
A17622.22
A17622.14
A17622.06
A17622.22EA
249
EUR
A17622.22
A17622.14
A17622.06
Adenin Hydrochlorid, (max. 5% H₂O) ≥98%
Adenin Hydrochlorid
Purity: 98+%
CAS No.: 2922-28-3
Molecular Formula: C5H5N5.HCl
Formula Weight: 171.59
Melting Point: ca 290° dec.
Beilstein Reference: 4009585
Formel:
C₅H₆ClN₅ Molecular Weight: 171.59 g/mol Schmelzpunkt: ∼ 290 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150864 CAS-Nummer: 2922-28-3 EINECS: 220-867-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...