Adenin Hydrochlorid, (max. 5% H₂O) ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Purin-6-amin Hydrochlorid, 1H-Adenin Hydrochlorid, 9H-Purin-6-amin Hydrochlorid, 6-Aminopurin Hydrochlorid

A17622.22 A17622.14 A17622.06
A17622.22EA 249 EUR
A17622.22 A17622.14 A17622.06
Adenin Hydrochlorid, (max. 5% H₂O) ≥98%
Adenin Hydrochlorid

Purity: 98+% CAS No.: 2922-28-3 Molecular Formula: C5H5N5.HCl Formula Weight: 171.59 Melting Point: ca 290° dec. Beilstein Reference: 4009585

Formel: C₅H₆ClN₅
Molecular Weight: 171.59 g/mol
Schmelzpunkt: ∼ 290 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150864
CAS-Nummer: 2922-28-3
EINECS: 220-867-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR