L(+)-Histidin Hydrochlorid Monohydrat ≥99%

Lieferant: Thermo Fisher Scientific

A17627.18 A17627.30 A17627.36
A17627.18EA 43.5 EUR
A17627.18 A17627.30 A17627.36
L(+)-Histidin Hydrochlorid Monohydrat ≥99%
L(+)-Histidin Hydrochlorid Monohydrat

Purity: 99% CAS No.: 5934-29-2 Molecular Formula: C6H9N3O2.HCl.H2O Formula Weight: 209.63 (191.62anhy) Melting Point: ca 240° dec. Beilstein Reference: 4168261 Merck Reference: 14,4720 Optical Rotation: +10° (c=5 in 5N HCl) MDL No.: MFCD00151027

Formel: C₆H₉N₃O₂·HCl·H₂O
Dichte: 1,485 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00151027
CAS-Nummer: 5934-29-2
EINECS: 211-438-9
Merck Index: 14,4720
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=561d2381f930afe618f78d680790ea9a43f6596c&vid=8aacf6dc4ef5382410cb743498272b48c3c36999&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR