Natriumthiosulfat, anhydrous 99%

Lieferant: Thermo Fisher Scientific

Synonyme: di-Natriumthiosulfat, Antichlor, Fixiersalz, Natriumhyposulfit

A17629.0E A17629.36
A17629.0EEA 106 EUR
A17629.0E A17629.36
Natriumthiosulfat, anhydrous 99%
Natriumthiosulfat

Purity: 99% CAS No.: 7772-98-7 Molecular Formula: Na2O3S2 Formula Weight: 158.10 Odour: Odourless Merck Reference: 14,8694 MDL No.: MFCD00003499

Formel: Na₂S₂O₃
Molecular Weight: 158.11 g/mol
Siedepunkt: 100
Schmelzpunkt: 48
Dichte: 1.667
Lagertemperatur: Raumtemperatur
MDL: MFCD00003499
CAS-Nummer: 7772-98-7
EINECS: 231-867-5
Merck Index: 14,8694

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR