2,2'-Dibrombiphenyl (PBB Nr. 4) ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 1-Brom-2-(2-bromphenyl)benzol

A17632.03 A17632.06 A17632.14
A17632.03EA 57.5 EUR
A17632.03 A17632.06 A17632.14
2,2'-Dibrombiphenyl (PBB Nr. 4) ≥98%
2,2'-Dibrombiphenyl (PBB Nr. 4)

Purity: 98% CAS No.: 13029-09-9 Molecular Formula: C12H8Br2 Formula Weight: 312.02 Melting Point: 78-81° UN No.: 3152 MDL No.: MFCD00093707

Formel: C₁₂H₈Br₂
Molecular Weight: 312 g/mol
Schmelzpunkt: 78…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00093707
CAS-Nummer: 13029-09-9
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR