2,2'-Dibrombiphenyl (PBB Nr. 4) ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
1-Brom-2-(2-bromphenyl)benzol
A17632.03
A17632.06
A17632.14
A17632.03EA
57.5
EUR
A17632.03
A17632.06
A17632.14
2,2'-Dibrombiphenyl (PBB Nr. 4) ≥98%
2,2'-Dibrombiphenyl (PBB Nr. 4)
Purity: 98%
CAS No.: 13029-09-9
Molecular Formula: C12H8Br2
Formula Weight: 312.02
Melting Point: 78-81°
UN No.: 3152
MDL No.: MFCD00093707
Formel:
C₁₂H₈Br₂ Molecular Weight: 312 g/mol Schmelzpunkt: 78…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00093707 CAS-Nummer: 13029-09-9 ADR: 9 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...