3,6-Dithia-1,8-octandiol ≥97%

Lieferant: Thermo Fisher Scientific

A17654.09 A17654.18 A17654.30
A17654.09EA 23.1 EUR
A17654.09 A17654.18 A17654.30
3,6-Dithia-1,8-octandiol ≥97%
3,6-Dithia-1,8-octandiol

Purity: 97% CAS No.: 5244-34-8 Molecular Formula: HOCH2CH2SCH2CH2SCH2CH2OH Formula Weight: 182.31 Melting Point: 63-67° Boiling Point: 170°/0.8mm MDL No.: MFCD00002911

Formel: C₆H₁₄O₂S₂
Siedepunkt: 170 °C (0,8 mmHg)
Schmelzpunkt: 63…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002911
CAS-Nummer: 5244-34-8
EINECS: 226-045-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR