3,6-Dithia-1,8-octandiol ≥97%
Lieferant: Thermo Fisher Scientific
A17654.09
A17654.18
A17654.30
A17654.09EA
23.1
EUR
A17654.09
A17654.18
A17654.30
3,6-Dithia-1,8-octandiol ≥97%
3,6-Dithia-1,8-octandiol
Purity: 97%
CAS No.: 5244-34-8
Molecular Formula: HOCH2CH2SCH2CH2SCH2CH2OH
Formula Weight: 182.31
Melting Point: 63-67°
Boiling Point: 170°/0.8mm
MDL No.: MFCD00002911
Formel:
C₆H₁₄O₂S₂ Siedepunkt: 170 °C (0,8 mmHg) Schmelzpunkt: 63…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002911 CAS-Nummer: 5244-34-8 EINECS: 226-045-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...