3-Nitrobenzolsulfonylchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-Nitrobenzolsulfochlorid, 3-Nitrobenzolsulfonsäurechlorid, 3-Nitrobenzol-1-sulfonylchlorid

A17670.22
A17670.22EA 234 EUR
A17670.22
3-Nitrobenzolsulfonylchlorid ≥98%
3-Nitrobenzolsulfonylchlorid

Purity: 98% CAS No.: 121-51-7 Molecular Formula: C6H4ClNO4S Formula Weight: 221.62 Flash Point: 110°(230°F) Melting Point: 59-63° Beilstein Reference: 746542 UN No.: 3261 MDL No.: MFCD00007435

Formel: C₆H₄ClNO₄S
Molecular Weight: 221,62 g/mol
Schmelzpunkt: 59…63 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007435
CAS-Nummer: 121-51-7
EINECS: 204-476-2
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR