Zellulose, mikrokristallin
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Zellulose
A17730.36
A17730.0C
A17730.0E
A17730.36EA
71.3
EUR
A17730.36
A17730.0C
A17730.0E
Zellulose, mikrokristallin
Zellulose
CAS No.: 9004-34-6
Physical Form: Powder
Appearance: White
Merck Reference: 14,1965
MDL No.: MFCD00081512
Formel:
(C₁₂H₂₂O₁₁)n Schmelzpunkt: 260…270 °C Dichte: 1,27…1.6 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081512 CAS-Nummer: 9004-34-6 EINECS: 232-674-9 Merck Index: 14,1965 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...