5-Methyl-2-thenoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Methylthiophene-2-carboxylic acid

A17737.06 A17737.14
A17737.06EA 46.9 EUR
A17737.06 A17737.14
5-Methyl-2-thenoesäure ≥98%
5-Methyl-2-thenoesäure

Purity: 98+% CAS No.: 1918-79-2 Molecular Formula: C6H6O2S Formula Weight: 142.18 Melting Point: 136-138° Beilstein Reference: 113857

Formel: C₆H₆O₂S
Schmelzpunkt: 136…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005439
CAS-Nummer: 1918-79-2
EINECS: 217-640-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR