Bromthymolblau

Lieferant: Thermo Fisher Scientific

A17746.09 A17746.18 A17746.30
A17746.09EA 34.8 EUR
A17746.09 A17746.18 A17746.30
Bromthymolblau
Bromthymolblau

CAS No.: 76-59-5 Molecular Formula: C27H28Br2O5S Formula Weight: 624.39 Melting Point: 200-202° Beilstein Reference: 373934 Merck Reference: 14,1445 MDL No.: MFCD00005872

Formel: C₂₇H₂₈Br₂O₅S
Schmelzpunkt: 200…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005872
CAS-Nummer: 76-59-5
EINECS: 200-971-2
Merck Index: 14,1445

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR