Benzo[h]chinolin 98%

Lieferant: Thermo Fisher Scientific

A17794.03 A17794.06
A17794.03EA 28 EUR
A17794.03 A17794.06
Benzo[h]chinolin 98%
Benzo[h]chinolin

Purity: 98% CAS No.: 230-27-3 Molecular Formula: C13H9N Formula Weight: 179.22 Flash Point: 110°(230°F) Melting Point: 48-52° Boiling Point: 233°/47mm Beilstein Reference: 120249 MDL No.: MFCD00004984

Formel: C₁₃H₉N
Siedepunkt: 233 °C (47 mmHg)
Schmelzpunkt: 48…52 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004984
CAS-Nummer: 230-27-3
EINECS: 205-937-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR