Natriumformiat ≥98%

Lieferant: Thermo Fisher Scientific

A17813.0B A17813.0I A17813.30
A17813.0BEA 33.9 EUR
A17813.0B A17813.0I A17813.30
Natriumformiat ≥98%
Natriumformiat

Purity: 98% CAS No.: 141-53-7 Molecular Formula: HCO2Na Formula Weight: 68.01 Melting Point: 259-262° Beilstein Reference: 3595134 Merck Reference: 14,8621 MDL No.: MFCD00013101

Formel: HCO₂Na
Schmelzpunkt: 259…262 °C
Dichte: 1,92
Lagertemperatur: Raumtemperatur
MDL: MFCD00013101
CAS-Nummer: 141-53-7
EINECS: 205-488-0
REACH: 01-2119486468-21
Merck Index: 14,8621

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR