5-Brom-1,3-benzodioxol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Bromo-1,2-(methylenedioxy)benzene, 5-Bromobenzo[d][1,3]dioxole

A17906.06 A17906.14 A17906.22 A17906.36
A17906.06EA 26 EUR
A17906.06 A17906.14 A17906.22 A17906.36
5-Brom-1,3-benzodioxol ≥98%
5-Brom-1,3-benzodioxol

Purity: 98% CAS No.: 2635-13-4 Molecular Formula: C7H5BrO2 Formula Weight: 201.02 Flash Point: 110°(230°F) Refractive Index: 1.5840 Boiling Point: 228° Beilstein Reference: 127407 MDL No.: MFCD00005821

Formel: C₇H₅BrO₂
Molecular Weight: 201.02 g/mol
Siedepunkt: 228 °C
Dichte: 1.669
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005821
CAS-Nummer: 2635-13-4
EINECS: 220-123-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR