4,5-Dichlor-2-nitroanilin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
A17908.06
A17908.06EA
43.3
EUR
A17908.06
4,5-Dichlor-2-nitroanilin ≥98%
4,5-Dichlor-2-nitroanilin
Purity: 98%
CAS No.: 6641-64-1
Molecular Formula: C6H4Cl2N2O2
Formula Weight: 207.02
Melting Point: 180-182°
Beilstein Reference: 643127
Formel:
C₆H₄Cl₂N₂O₂ Molecular Weight: 207,02 g/mol Schmelzpunkt: 180…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007770 CAS-Nummer: 6641-64-1 EINECS: 229-657-3 UN: 2237 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...