Hyperosid (aus Hypericum perforatum) ≥97,0% (durch HPLC), rein
Lieferant: PanReac AppliChem
|
Achtung
|
Synonyme:
Hyperoside, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one, Hyperin, Quercetin-3-galactoside
A1791.0100
A1791.0100EA
494
EUR
A1791.0100
Hyperosid (aus Hypericum perforatum) ≥97,0% (durch HPLC), rein
Hyperosid
Originated from hypericum perforatum.
Formel:
C₂₁H₂₀O₁₂ Molecular Weight: 464,38 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00016933 CAS-Nummer: 482-36-0 |
Spezifikation Testergebnisse
State of Matter | Solid |
Storage Temperature | 2 - 8 °C |
Assay (HPLC) | min. 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...