Bromthymolblau Natriumsalz

Lieferant: Thermo Fisher Scientific

A17913.09 A17913.18
A17913.09EA 39 EUR
A17913.09 A17913.18
Bromthymolblau Natriumsalz
Bromthymolblau Natriumsalz

CAS No.: 34722-90-2 Molecular Formula: C27H27Br2NaO5S Formula Weight: 646.38 Melting Point: 200-202° MDL No.: MFCD00077263

Formel: C₂₇H₂₇Br₂NaO₅S
Schmelzpunkt: 200…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00077263
CAS-Nummer: 34722-90-2
EINECS: 252-169-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR