Lithiumacetat Dihydrat ≥99%
Lieferant: Thermo Fisher Scientific
A17921.0B
A17921.30
A17921.0BEA
185
EUR
A17921.0B
A17921.30
Lithiumacetat Dihydrat ≥99%
Lithiumacetat Dihydrat
Purity: 99%
CAS No.: 6108-17-4
Molecular Formula: LiOOCCH3.2H2O
Formula Weight: 102.01 (65.98anhy)
Melting Point: 54-57°
Beilstein Reference: 3564320
Merck Reference: 14,5521
MDL No.: MFCD00066949
Formel:
H₃CCOOLi·2H₂O Schmelzpunkt: 54…57 °C Dichte: 1,300 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066949 CAS-Nummer: 6108-17-4 EINECS: 208-914-3 Merck Index: 14,5521 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...