Polyethylenglycol 20.000
Lieferant: Thermo Fisher Scientific
CAS No.: 25322-68-3
Molecular Formula: H(OCH2CH2)nOH
Flash Point: 260°(500°F)
Physical Form: Powder or flake
Density: 1.20
Appearance: White
Odour: Odourless
pH: 5-7
Melting Point: 63-66°
Merck Reference: 14,7568
MDL No.: MFCD00081839
Formel:
H(OCH₂CH₂)nOH Schmelzpunkt: 63…66 °C Dichte: 1,20 Flammpunkt: 260 °C (500 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081839 CAS-Nummer: 25322-68-3 EINECS: 203-473-3 Merck Index: 14,7568 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...