D(-)-Diethyltartrat ≥99%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
(-)-Diethyl-D-tartrat, D(-)-Threarsäurediethylester, (S,S)-(-)-2,3-Dihydroxy-1,4-butandisäurediethylester, D(-)-Weinsäurediethylester, (S,S)-(-)-2,3-Dihydroxybutandisäurediethylester, (S,S)-(-)-Weinsäurediethylester, (S,S)-(-)-Diethyltartrat, (S,S)-(-)-2,3-Dihydroxybernsteinsäurediethylester
Purity: 99%
CAS No.: 13811-71-7
Molecular Formula: C8H14O6
Formula Weight: 206.19
Flash Point: 93°(199°F)
Refractive Index: 1.4460
Melting Point: 17°
Boiling Point: 280°
Beilstein Reference: 1727143
Optical Rotation: -26.5° (c=1 in water)
MDL No.: MFCD00064451
Formel:
C₈H₁₄O₆ Siedepunkt: 280 °C Schmelzpunkt: 17 °C Dichte: 1,205 Flammpunkt: 93 °C (199 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064451 CAS-Nummer: 13811-71-7 EINECS: 237-458-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...