D-Cycloserin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
D(+)-Cycloserin, D-4-Aminoisoxazolidin-3-on, (R)-(+)-Cycloserin
A18000.03
A18000.06
A18000.03EA
71.4
EUR
A18000.03
A18000.06
D-Cycloserin ≥98%
D-Cycloserin
Purity: 98+%
CAS No.: 68-41-7
Molecular Formula: C3H6N2O2
Formula Weight: 102.09
Storage Temperature: 2°C to 8°C
Melting Point: 137° dec.
Beilstein Reference: 80798
Merck Reference: 14,2751
Optical Rotation: +115° (c=2 in water)
MDL No.: MFCD00005353
Formel:
C₃H₆N₂O₂ Molecular Weight: 102.09 g/mol Schmelzpunkt: 137 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00005353 CAS-Nummer: 68-41-7 EINECS: 200-688-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...