Oxalyldichlorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A18012.0B
A18012.18
A18012.22
A18012.30
A18012.0BEA
403
EUR
A18012.0B
A18012.18
A18012.22
A18012.30
Oxalyldichlorid ≥98%
Oxalyldichlorid
Purity: 98%
CAS No.: 79-37-8
Molecular Formula: C2Cl2O2
Formula Weight: 126.93
Physical Form: Liquid
Density: 1.455
Refractive Index: 1.4290
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Acrid
Melting Point: -12°
Boiling Point: 63-64°
Beilstein Reference: 1361988
Merck Reference: 14,6914
UN No.: 2927
MDL No.: MFCD00000704
Formel:
C₂Cl₂O₂ Siedepunkt: 63…64 °C Schmelzpunkt: –12 °C Dichte: 1,455 Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000704 CAS-Nummer: 79-37-8 EINECS: 201-200-2 UN: 2927 ADR: 6.1,II Merck Index: 14,6914 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...