m-Kresolpurpur Natriumsalz

Lieferant: Thermo Fisher Scientific

A18025.06 A18025.14
A18025.06EA 53.1 EUR
A18025.06 A18025.14
m-Kresolpurpur Natriumsalz
m-Kresolpurpur Natriumsalz

CAS No.: 62625-31-4 Molecular Formula: C21H17NaO5S Formula Weight: 404.42 MDL No.: MFCD00010177

Formel: C₂₁H₁₇NaO₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD00010177
CAS-Nummer: 62625-31-4
EINECS: 263-656-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR