5-Sulfoisophthalsäure Natriumsalz 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A18028.22
A18028.30
A18028.36
A18028.22EA
55.7
EUR
A18028.22
A18028.30
A18028.36
5-Sulfoisophthalsäure Natriumsalz 95%
5-Sulfoisophthalsäure Natriumsalz
Purity: 95%
CAS No.: 6362-79-4
Molecular Formula: C8H5NaO7S
Formula Weight: 268.18
Melting Point:
Formel:
C₈H₆O₇S Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007495 CAS-Nummer: 6362-79-4 EINECS: 228-845-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...