Europiumtriacetat Hydrat 99%

Lieferant: Thermo Fisher Scientific

A18069.03
A18069.03EA 38.3 EUR
A18069.03
Europiumtriacetat Hydrat 99%
Europiumtriacetat Hydrat

Purity: 99.99% CAS No.: 62667-64-5 Molecular Formula: Eu(OOCCH3)3.xH2O Formula Weight: 329.10(anhy) MDL No.: MFCD00150115

Formel: C₆H₉EuO₆·H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00150115
CAS-Nummer: 62667-64-5
EINECS: 214-670-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR