2-Methoxyphenylisocyanat 98

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-Isocyanato-2-methoxybenzene

A18075.06 A18075.14
A18075.06EA 44.3 EUR
A18075.06 A18075.14
2-Methoxyphenylisocyanat 98
2-Methoxyphenylisocyanat

Purity: 98% CAS No.: 700-87-8 Molecular Formula: C8H7NO2 Formula Weight: 149.15 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5414 Odour: Pungent Boiling Point: 94-96°/10mm Beilstein Reference: 387700 UN No.: 2206 MDL No.: MFCD00002004

Formel: C₈H₇NO₂
Molecular Weight: 149.15 g/mol
Siedepunkt: 94…96 °C (10 mmHg)
Dichte: 1.122
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00002004
CAS-Nummer: 700-87-8
EINECS: 211-853-5
UN: 2206
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR