2,6-Difluorbenzaldehyd ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A18099.14
A18099.06
A18099.14EA
217
EUR
A18099.14
A18099.06
2,6-Difluorbenzaldehyd ≥97%
2,6-Difluorobenzaldehyde
Purity: 97%
CAS No.: 437-81-0
Molecular Formula: C7H4F2O
Formula Weight: 142.11
Flash Point: 73°(163°F)
Refractive Index: 1.5070
Melting Point: 15-17°
Boiling Point: 85-86°/20mm
Beilstein Reference: 1935273
MDL No.: MFCD00010293
Formel:
C₇H₄F₂O Siedepunkt: 85…86 °C (20 mmHg) Schmelzpunkt: 15…17 °C Dichte: 1,317 Flammpunkt: 73 °C (163 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010293 CAS-Nummer: 437-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...